Vikan® 16" UST Push Broom Head, Medium Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
The 16" Ultra Safe Technology (UST) medium push broom is effective for sweeping in dry or wet high risk areas.
- Polypropylene block, polyester bristles
- Euro (hygienic) thread type
The angled arrangement of the Bristle Security Units ensures efficient and effective cleaning, while the lack of resin, glue, or staples means bristle units are easy to clean, FDA-compliant, and more secure.
Broom head dimensions: 15.25L×1.875W×4.7"H
Customers Who Bought This Also Bought