Vikan® 33" Aluminum Handle, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
Ideal for use when cleaning tubes and drains with tube brushes.
- The handle fits most Remco or Vikan® products
- Aluminum construction with color-coded polypropylene grips
- Euro (hygienic) thread type
Also suitable for use as a broom handle.
Customers Who Bought This Also Bought