Vikan® Hygienic Wall Bracket, Grip Band Module, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
Grip band module is designed for storing cleaning tools without a hanging hole.
- Can hold products with a diameter of 1.1" to 1.3"
- Polypropylene/TPE rubber/polyamide construction
The grip band module is easy to disassemble for cleaning or replacement.
Customers Who Bought This Also Bought