Vikan® Hygienic Hi-Flex Wall Bracket System, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
Provides a flexible solution for storing cleaning tools and utensils in any area of the worksite.
- Can hold products weighing up to 6.61 lbs.
- Supplied with three grip band modules and two hooks
- Polypropylene/TPE rubber/polyamide construction
Modules and rails are easy to disassemble for cleaning or replacement.
Customers Who Bought This Also Bought