Vikan® Combo Push Broom Heads, Soft/Stiff Bristles, 16", Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
Fully color-coded sweeping broom head features two types of bristles to pick up every particle.
- Polypropylene block, polyester bristles
- Euro (hygienic) thread type
Stiff bristles loosen stubborn dirt, and soft bristles sweep away loose particles.
Broom head dimensions: 16.1L×2.6W×4.2"H
Customers Who Bought This Also Bought