BenchBin™ Benchtop Biohazard Bin
Supplier: MTC BIO, INC.
Certificates
About this item
The BenchBin™ is a compact, convenient, and safe biohazard bin for the benchtop or workstation.
- Safely dispose of tips, tubes, gloves and more
- Secures and covers hazardous waste
- Easy push open/close spring lid
- Compact footprint (7×5×7")
For the temporary storage of biohazardous material during a lab activity or assay that doesn't require multiple trips to the biohazard bin. The sleek and compact design requires minimal space on the lab bench while the closed-lid system and easy bag removal creates a safe and convenient solution for waste .
Ordering information: Red biohazard bags (8.5×11") are purchased separately in packs of 100.
Delivery: The BenchBin™ Starter Kit includes one BenchBin and 400 red 8.5×11" autoclave/ biohazard bags.
Customers Who Bought This Also Bought