VWR® Freezer Racks for Covid-19 Vaccine Box Storage
Fournisseur: CRYSTAL TECHNOLOGY & INDUSTRIES
Certificats
A propos de cet article
Create an organized inventory control system. Economical smart storage designed for Pfizer and Moderna Covid-19 vaccine box storage. An ideal way to keep vaccines organized and ready for easy retrieval.
- Constructed from pharmaceutical grade stainless steel
- Double welds for added strength and long life
- Smoothed edges for easy handling
- Variety of freezer rack choices
- Make vaccine storage and retrieval fast and efficient
- Lifetime warranty
Specially designed inventory control freezer racks to make storage and retrieval of Pfizer and Moderna Covid-19 vaccine boxes more efficient. Choose between drawer style or side access, or even a convenient vertical rack to maximize cold storage space.
Delivery: Each rack comes with a lifetime warranty.
Customers Who Bought This Also Bought