Medicor® Oral and Nasal Swab Simulator
Fournisseur: ELEVATE HEALTHCARE, INC. SE
Certificats
A propos de cet article
Sample collection training.
- Available in three skin tones
- Practice both oral and nasal swab collection
The advent of the COVID-19 virus highlights the need for ensuring students are properly trained in collecting samples. Made entirely of proprietary MedicSkin and ideal for students to practice how to properly perform a nasal or oral swab, this simulator features true-to-life anatomy, including the tongue, uvula and teeth, providing a realistic training experience.
Customers Who Bought This Also Bought