Medicor® Sticky Wounds
Fournisseur: ELEVATE HEALTHCARE, INC. SE
Certificats
A propos de cet article
Add to manikins or standard patients.
- Easy to remove and reuse
- Expand training with existing manikins
- Skin tone colors match Medicor and CAE
Medicor sticky wounds are made entirely of MedicGel for a lifelike appearance. Able to stick to any surface without adhesive, these wounds can be applied to any manikin or standardized patient. All sticky wounds are stain-resistant and hypoallergenic.
TIP: If the product is not sticking well due to dust or other contaminants, rinse with tap water to reactivate the material.
Customers Who Bought This Also Bought