Medicor® Injectable Abdomen
Fournisseur: ELEVATE HEALTHCARE, INC. SE
Certificats
A propos de cet article
Practice subcutaneous injection.
- Available in three skin tones
- Available in a belted version
Made entirely of proprietary MedicSkin, injectable abdominal models provide a lifelike experience to practice administering subcutaneous injections. Students get a realistic haptic experience squeezing the tissue and placing the injection. The interior of the model contains a sponge, making it ideal to practice injecting fluid into the model without deterioration or molding. Choose from the base model or the belted model with protection plate. The protection plate provides a barrier to avoid accidental punctures while the belt allows learners to securely attach to manikins or standardized patients.
Customers Who Bought This Also Bought