Medicor® Peripheral Intravenous (IV) Catheterization Hand
Fournisseur: ELEVATE HEALTHCARE, INC. SE
Certificats
A propos de cet article
Easily practice small vein phlebotomy.
- Available In three skin tones
- Mold resistant
- Self healing skin to 22G
Made entirely of proprietary MedicSkin, this lifelike hand provides students with a realistic experience for practicing peripheral IV insertion and phlebotomy procedures in the small veins of the hand. Students can practice on the dorsal, basilic, or cephalic veins. Track marks heal quickly, allowing students to practice repeatedly.
Ordering information: Skin and veins are not replaceable.
Customers Who Bought This Also Bought