VACUUBRAND® MD 1C Chemistry Diaphragm Vacuum Pump, BrandTech®
# de catalogue 76529-726
Fournisseur: Brandtech
Spécifications
- Description:MD 1C vacuum pump
- Environmentally Preferable:Energy Efficient,Waste Reducing,Water Efficient
- Cat. no.:76529-726
- Electrical:120/230 V, 50/60 Hz (US plug)
- Ultimate Vacuum:2 mbar (1.5 torr)
- Free Air Capacity:25 L/min (0.9 CFM)
Spécifications
A propos de cet article
Powerful 2 millibar performance for evaporation of high boiling point solvents.
- Oil-free
- Corrosion-resistant fluoropolymer/borosilicate construction
- Whisper-quiet
- Ultra-low maintenance (>15000 service hours)
- Compact footprint
The VACUUBRAND MD 1C chemistry diaphragm vacuum pump provides high flow rates at working vacuum levels to evaporate high boiling point solvents. A three-stage design, with integrated gas ballast provides a high tolerance for condensible vapors. A full fluropolymer flowpath provides superior resistance to chemicals. Pump is oil-free, with typical service intervals in excess of 15000 hours of operation (15 months of 24/7 operation). Suitable for many benchtop applications, such as rotary evaporation, gel drying, and small vacuum ovens.
Certifications: cTŰVus
Ordering information: Two year warranty.
Delivery: Supplied with operating manual and US plug.
Packaging: Recyclable packaging.
Customers Who Bought This Also Bought