Cole-Parmer Stuart Hybrigene and Hybridizer Incubators, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Use and recover volumes as low as 5 ml, ensuring preservation of precious samples.
- Radioactive - safe protective casing and non-drip tube design minimize risk to user
- Temperature resolution: 0.1 °C
With excellent stability of ±0.1 °C within the tubes, these Hybrigene and Hybridizer incubators are great for nucleic hybridization applications such as Northern, Southern, and Western blots. Each incubator features two shakers - rotisserie shaker with rotational speed from 0 to 20 rpm and platform shaker with rocking speed up to 60 oscillation/minute. A removable drip tray provides for easy cleaning. Adjustable feet make leveling on uneven surfaces quick and easy.
Ordering information: 76531-222 and 76531-226 are supplied with tubes. However 76531-224 is not supplied with tubes and these need to be ordered separately.
Spécifications
- Catalog No:
- 76531-226
- 76531-222
- 76531-224
- Temperature accuracy:
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- Temperature stability:
- Chamber: 1.0 °C; tube: ±0.1 °C
- Chamber: 1.0 °C; tube: ±0.1 °C
- Chamber: 1.0 °C; tube: ±0.1 °C
- Display resolution:
- LED
- LED
- LED
- Power:
- 750 W
- 750 W
- 750 W
- Shaking speed:
- 0 - 20 rpm
- 0 - 20 rpm
- 0 - 20 rpm
- Electrical:
- 120 VAC
- 120 VAC
- 120 VAC
- Dimensions:
- 385L×355W×385H mm (15×14×15")
- 285L×385W×555H mm (11.25×15×21")
- 385L×355W×385H mm (15×14×15")