About this item
These reliable, easy-to-wear gloves are more economical than standard nitrile gloves and are suited for a wide variety of tasks.
- Ambidextrous and powder-free
- Beaded cuffs
- 3.2 mil thickness
- Superior quality, AQL 1.5 (exceeds the ASTM D 5151 standard for critical defects or pinholes)
- Not made with natural rubber latex
- Waste less with the one-at-a-time dispenser box
- 250% more gloves per case (versus standard nitrile gloves)
The search for a comfortable, protective exam glove with exceptional value is over, thanks to Kimtech™ Element™ nitrile examination gloves. These gloves deliver enhanced dexterity, thanks to textured fingertips, and are not made with natural rubber latex. The gloves are static dissipative in use.
They are packaged in an innovative dispenser box, which incorporates two separate openings. The first, smaller opening is used when the box is full and helps control dispensing to lessen waste from dropped gloves or multiple dispensing. When the box is half empty, the second, larger opening allows easier access to the gloves. They be an important piece for creating a more efficient and responsible workplace.
Kimtech™ Element™ nitrile gloves can be recycled with the Kimberly-Clark Professional RightCycle program, the first large-scale recycling program for nonhazardous lab and cleanroom waste.
Specifications
- Catalog No:
- 76520-544
- 76520-538
- 76520-536
- 76520-542
- 76520-540
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile