Kimtech™ G3 EvT Prime™ Nitrile Gloves, Kimberly-Clark
Fournisseur: Kimberly-Clark
Certificats
A propos de cet article
White nitrile cleanroom gloves.
- Non sterile, ambidextrous glove with textured fingertips
- 4.72 mil / 0.12 mm thickness
- Beaded cuff
- Amide-free, powder-free
These gloves are double chlorinated and double bagged. These gloves are not made with silicone.
Recyclable via the RightCycle™ program.
Spécifications
- Catalog No:
- 76501-046
- 76501-048
- 76501-050
- 76501-052
- 76501-054
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile