VWR®, Duster Spray
# de catalogue 76528-070
Fournisseur: VWR International
Spécifications
- Volume:284 g (10 oz.)
- Description:VWR® Duster spray
Spécifications
A propos de cet article
Easily remove debris from hard-to-reach places.
- All-purpose sterilized cleaner is nonflammable
- Remove dust, dirt, and microscopic debris from home and office equipment
- Safe for the ozone with ultra-low global warming potential (GWP)
VWR® air duster spray removes dust, dirt, and debris from delicate or hard-to-reach places such as keyboards, copiers, lab equipment, fans, and more. The aerosol cleaning spray provides jets of cleaning power that is easy and safe to use.
Delivery: Extension tube.
Customers Who Bought This Also Bought