ParafiTAG™ Xylene Resistant Labels for Paraffin Wax Blocks
# de catalogue 76210-654
Fournisseur: GA INTERNATIONAL INC
Spécifications
- Color:White
- Cat. no.:76210-654
- Packaging:4000/roll
- Core Size:1" (25 mm)
- Dimensions:0.75×0.25" (19.1×6.4 mm)
Spécifications
A propos de cet article
These thermal transfer printable thermoplastic labels adhere strongly to paraffin wax blocks and other hard to stick surfaces such as Teflon, Polytetrafluoroethylene (PTFE), Perfluoroalkoxy (PFA), parchment paper, and more.
- Adheres to difficult surfaces, including paraffin wax blocks
- Withstands exposure to harsh chemicals and solvents
- Printable with all major barcode printers
- Perforations every two labels
- Temperature range: -55 to 70° C (-67 to 158° F)
ParafiTAG™ labels are water- and chemical- resistant, ideal for the identification of blocks of paraffin wax for histology applications. A chemical-resistant printout is obtained when printed with GA International XAR class solvent-resistant ribbons. The labels and printed information will resist xylene, toluene, alcohol, and other solvents and chemical exposure without additional protection.
Customers Who Bought This Also Bought