Classroom Chart Stand with Storage Bins, Luxor
# de catalogue 76519-964
Fournisseur: Luxor
Spécifications
- Color:Grey/green
- Description:Classroom whiteboard stand with storage bins
- Cat. no.:76519-964
- Dimensions:W×D: 31×25"
Height: 61 - 72"
Spécifications
A propos de cet article
Double-sided magnetic whiteboard with storage bins.
- Space-saving vertical orientation
- Chart hooks display easel paper pads, pocket charts and posters
- Painted steel surface resists ghosting and provides excellent erasability
- Great mobility with four casters, two with locking brakes
- Marker ledge holds books and stores whiteboard materials
Two durable plastic open bins hold supplies while handles allow for easy toting.
Customers Who Bought This Also Bought