A propos de cet article
Globe Glass™ graduated cylinders are calibrated to ASTM E1272 standards for precise measurement of volumes of liquids. A solid hexagonal base provides stability, and an included polyethylene (PE) bumper guard helps to protect against accidental breakage. These graduated cylinders are calibrated TD, 'To Deliver', and are available in sizes ranging from 5 to 2000 ml, with Class A or Class B calibration tolerance options. Class A graduated cylinders include a batch calibration certificate.
- Calibrated TD; 'To Deliver'
- Class A calibration tolerance for highest accuracy and the most critical laboratory work
- Class B calibration tolerance for general laboratory work
- Pour spout for easy liquid transfer
- Beaded rim for strength and durability
- High resistance to chemicals
- Dual scale metric graduations; durable white enamel printing
Globe Glass™ lab glassware is made exclusively from ASTM E438, Type I, Class A, 3.3 borosilicate glass for excellent resistance to thermal shock and high chemical resistance to most acids, alkalis, and solvents. Globe Glass™ lab glassware meets applicable ASTM standards for volume, shape, dimensions, graduation markings, and calibration. These ASTM standards are clearly marked for easy confirmation that each piece of laboratory glassware will meet the needs of laboratory professionals. Computer controlled, automated production lines ensure that this laboratory glassware has consistent wall thickness, flat bottoms free of striations, and precise forming of rims and spouts.
Permanent white enamel markings are ASTM compliant and easy to read. Dual metric scale graduations are included on most items for easy reference while filling or dispensing. A large, convenient marking area is provided for coding or identification. Globe Glass™ laboratory glassware is manufactured under stringent quality controls. Manufacturing facilities are ISO 9001 certified and in-house calibration is performed in an ISO 17025 certified laboratory.