A propos de cet article
Simple to use benchtop meter for easily measuring pH and ORP.
- With six instructional menu buttons, auto buffer recognition or auto temperature compensation, AB23 series is an easy and straightforward meter for measurement
- Large 5" bright LCD display with complete measurement information simplifies operation and allows you to monitor the results from a distance
- Featuring a compact stand-alone electrode holder, AB23 series increases the flexibility of experiments and fits ideally in any lab
Designed with a large LCD Display and six instructional menu buttons, the AB23 series bench meter is simple and straightforward. A 99-item memory for measurements ensures efficient data collection. A stand-alone electrode holder with a small base provides the user greater flexibility when running experiments and makes the AB23 benchtop meter a perfect space saving meter for your lab.
Also available is a USB-powered, low-profile stirrer which plugs directly into the pH meter. Includes stir bar.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 76518-264
- 76518-266
- mV range:
- –1999...1999
- –1999...1999
- mV resolution:
- 1
- 1
- mV accuracy:
- ±1 mV
- ±1 mV
- Temperature range:
- 0...100 °C
- 0...100 °C
- Temperature resolution:
- 0.1 °C
- 0.1 °C
- Display:
- LCD with backlight
- LCD with backlight
- Parameters:
- pH / ORP
- pH / ORP