Universal Pippette Tips, Racked, Sterile, Crystal Industries
Fournisseur: CRYSTAL TECHNOLOGY & INDUSTRIES
Certificats
A propos de cet article
Universal Pipette Tips designed to assure fit with all major brands of pipettors. Low retention tips reduce residue on the inner walls to ensure accurate results. Easy to use and store in reusable racks, color-coded by tip size.
- Produced in Class 100000 cleanroom
- Pharmaceutical grade polypropylene
- DNase-free, RNase-free, Pryogen-free, no endotoxins or PCR inhibitors
- Works with various pipettors
- Sterile (gamma radiated)
- Hydrophobic filter to avoid cross contamination
- Low retention tips for best accuracy
Premium materials and optimal design ensure these Universal Pippette Tips deliver great performance in the laboratory.
Packaging: 10 racks per pack.
Customers Who Bought This Also Bought