VWR® 4 ml Screw-Thread Vials, Kits, and Caps
Supplier: VWR International
Certificates
About this item
4 ml screw thread vials are designed for use with Waters® WISP® (48-position) and Shimadzu™ autosamplers.
- Vials are precision manufactured from high-purity borosilicate glass
- 13-425 Caps and vials matched for best closure integrity
- Available with or without writing patch
Vials and caps available as stand alone items or in a convenience kit configuration.
Specifications
- Catalog No:
- 46610-739A
- 82028-438A
- 66009-876A
- 66009-880A
- 82028-440A
- 66010-562A
- Kit:
- 66009-876A
- 66009-880A
- Component vial:
- 66010-562A
- 66010-562A
- Component cap:
- 46610-706A
- 46610-706A