A propos de cet article
Gas filter system for reduced risk of column damage, sensitivity loss, and instrument downtime.
- Delivery of clean gases for accurate analyses
- Fast, leak-free filter replacement for reduced downtime
- Economical, with immediate payback
- Highly sensitive filter indicators provide maximum instrument protection
The Agilent gas clean purification system delivers clean gases, reducing the risks of column damage, sensitivity loss, and instrument downtime. Inserting a gas clean purification system in the gas line immediately before the instrument inlet greatly reduces the level of impurities, thus improving trace analysis. Contaminants entering your GC column will also be reduced, which is critical for high-temperature analysis and essential for longer column lifetime.
Replacing the filters when they have reached absorption capacity ensures maximum protection of your GC columns and analytical hardware. The sensitive indicators change color, alerting you that the filter needs to be replaced. A gas clean purification system allows you to use 99.996% (4.6) pure helium instead of the more expensive 99.999% (5.0) or 99.9999% (6.0) grade, while still yielding high-quality analytical results. You can expect cost savings of 30%.