ContactUsLinkComponent
CrossLab CE and CE/MS Tools, Agilent Technologies
CrossLab CE and CE/MS Tools, Agilent Technologies
# de catalogue CA5183-4669
COLUMN CUTTER,CE/CECCAP
CrossLab CE and CE/MS Tools, Agilent Technologies
# de catalogue CA5183-4669
Spécifications
  • Description:
    CE column cutter with diamond blade
  • Cat. no.:
    CA5183-4669
  • Supplier No.:
    5183-4669
Spécifications
A propos de cet article
For fast, convenient, and reproducible preparation of detection windows on fused silica capillaries.
  • Reproducible preparation of detection windows on fused-silica capillaries
  • Polyimide coating is removed without compromising the inner polymeric coating
  • Tool features three glass blocks with fine grooves to control the size of windows
Agilent supplies CE tools such as the window etching tool, designed for fast, convenient, and reproducible preparation of detection windows on fused-silica capillaries. The polyimide coating is removed without destroying the inner polymeric coating. The tool contains three glass blocks with fine grooves, precisely controlling the size of the windows.
Customers Who Bought This Also Bought