VWR® Manual Crimping and Decapping Tools
Fournisseur: VWR International
Certificats
A propos de cet article
Screw-stop in crimping handle allows for adjustment of crimping pressure. Decrimping tools provide convenient method for crimp seal removal.
- Easy and convenient handling
- Durable, high quality construction
- Adjustable screw setting facilitates optimum crimping
Adjustable in crimping pressure limitation by screw in handle.
Customers Who Bought This Also Bought