VWR® 20 mm Clear Crimp Seal Headspace Vials and Caps
Fournisseur: VWR International
Certificats
A propos de cet article
These 20 ml crimp top headspace vials are made from high-purity borosilicate glass.
- The beveled finish provides a sturdy rim that presses into the septum and creates a more effective seal
- Available with a round or flat bottom
Vials are converted from glass tubing to provide uniform wall thickness and promote even heat distribution.
Ordering information: 20 mm crimp seals and headspace vials sold separately.
Customers Who Bought This Also Bought