VWR® 11 mm Crimp Top Vials, Kits, and Inserts, Standard Opening
Fournisseur: VWR International
Certificats
A propos de cet article
These 11 mm crimp top, standard opening vials have a maximum fill volume of 1.8 ml and are manufactured from high purity borosilicate glass.
- Aluminum crimp closures provide a secure leak-resistant seal
- Aluminum seals must be applied with a crimping tool
- Spring promotes alignment and ensures insert seals against septa to help guard against evaporation loss
Standard opening design requires inserts with a diameter of 5 mm.
Spécifications
- Catalog No:
- 82028-402A
- 66020-953A
- 82028-404A
- 89239-006A
- 89239-004A
- Kit:
- 89239-006A
- 89239-004A
- Component vial:
- 82028-404A
- 66010-539A
- Component cap:
- 46610-744A
- 46610-744A