VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
Certificates
About this item
11 mm crimp top, wide neck opening design, allows easy filling, requires inserts with a diameter of 6 mm.
- Aluminum crimp closures provide a secure leak-resistant seal
- Aluminum seals must be applied with a crimping tool
- Spring promotes alignment and ensures insert seals against septa to help guard against evaporation loss
Wide opening vials feature 40% larger neck opening than standard opening vials, for easier access and reduced needle damage.
Specifications
- Catalog No:
- 82028-408A
- 46610-742A
- 46610-740A
- 82028-410A
- 66009-864A
- 66009-850A
- 66010-539A
- 89239-008A
- Kit:
- 66009-864A
- 66009-850A
- 89239-008A
- Component vial:
- 66010-539A
- 66010-539A
- 46610-742A
- Component cap:
- 46610-744A
- 46610-744A
- 46610-744A