VWR® 11 mm Snap Ring Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
Certificates
About this item
11 mm snap ring vials can be used with snap caps or aluminum crimp seal closures.
- Manufactured from clear or amber borosilicate glass or polypropylene
- Vials feature a write-on patch with graduation for convenient sample identification
- Wide neck opening design, allows easy filling, requires inserts with a diameter of 6 mm
- Spring promotes alignment and ensures Insert seals against septa to help guard against evaporation loss
Vials have 40% wider openings than standard vials to reduce risk of needle damage.
Specifications
- Catalog No:
- 82028-412A
- 66009-868A
- 82028-414A
- 66009-822A
- 10803-890A
- 66009-872A
- 82028-416A
- Kit:
- 66009-868A
- 66009-872A
- Component vial:
- 66009-822A
- 66009-822A
- Component cap:
- 46610-718A
- 46610-720A