Accessories for Corepoint™ Scientific Blood Bank Refrigerators and Plasma Freezer Units, Horizon Scientific
Fournisseur: Horizon Scientific
Certificats
A propos de cet article
These additional accessories are designed for use with Corepoint™ Scientific blood bank refrigerators and plasma freezers.
- Factory fitted options available are:
- The anchor bracket kit is designed to add extra anchoring support
- The upper solid ballast kit is an optional kit to replace the standard upper glycol bottle
- The lower glycerol bottle kit is an optional kit to replace the standard lower solid ballast
The optional IQ/OQ/PQ add on for use with Corepoint™ Scientific blood bank refrigerators and plasma freezers should be ordered at the same time as the unit as it is shipped with the unit.
Spécifications
- Catalog No:
- 76520-788
- 76520-794
- 76520-786
- 76520-784
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient