ContactUsLinkComponent
Ward's® Diorite Gneiss
Ward's® Diorite Gneiss
# de catalogue 470331-548
Fournisseur:  Avantor
GNEISS DIORITE THIN SECTION CA
Ward's® Diorite Gneiss
# de catalogue 470331-548
Fournisseur:  Avantor
Spécifications
  • Description:
    Diorite gneiss thin section
  • Cat. no.:
    470331-548
Spécifications
A propos de cet article
When minerals, rocks, and fossils cannot be accurately identified by macroscopic observation and testing, thin section slides can reveal the composition and structure of the specimens on a microscopic level.
Thin section slides are precisely ground to a thickness of 0.03 mm across the entire specimen area. Mineral and rock thin sections are mounted on 27×46 mm microscope slides, and protected by a glass coverslip. All thin section slides are labeled for reference.
Customers Who Bought This Also Bought