SymLine® SwitchBox and 3-Way Ball Valve, SCAT
Supplier: SCAT AMERICAS, INC.
Certificates
About this item
Automatically divert waste liquids into the reserve canister.
- Switchbox compatible to all electronic S.C.A.T. Europe and SymLine® level sensors
- Also compatible with the Signalbox and Built-in signalbox
- Many more applications, not only for waste disposal of HPLC systems
Improved safety through increased collecting volume, the reserve canister protects against overflowing, even for a longer time period. For example, waste can be disposed of over the weekend without operator supervision and without having to empty or change the full container.
Customers Who Bought This Also Bought