SymLine® Desktop and Rear Mounts
Fournisseur: SCAT AMERICAS, INC.
Certificats
A propos de cet article
The filling units are mounted in space-saving fashion within the workspace. Available as a special HPLC connection, for direct connection to waste tubing.
- Desktop mount can be integrated into nearly all working areas
- Flat desktop mount and HPLC waste collector in one
- Rear wall mount uses available working area capacity to provide more space for laboratory samples and experimental setups
- Waste can be disposed directly in the fume cupboard
The desktop mount can be integrated into nearly all working areas. Whether used in conjunction with a work table or a fume cupboard, it always provides efficient transfer of liquid waste into connected waste systems. Thanks to integrated connections for capillaries and tubes, the flat desktop mount installation protrudes only slightly from the worktop.
Customers Who Bought This Also Bought