J.T.Baker® Robotic Tips Type DX
Fournisseur: Ritter, part of Avantor
Certificats
A propos de cet article
Suitable for use on Dynex DSX and DS2 instruments. Available in non-conductive version, without filter, in container rack.
- 108 tips in one optimized container with lid
- Especially developed for the automated sample handling
- Highest quality specifications for the compatibility and cleanliness
- Blue sample tips for visual tip identification
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Spécifications
- Catalog No:
- 49136-0000
- 49137-0000
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)