J.T.Baker® Plate, Dilution Strips SW
Catalog # 43001-0031
Supplier: Ritter, part of Avantor
Specifications
- Color:Transparent
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- Cat. no.:43001-0031
- Sterile:-
Specifications
About this item
Suitable for use on Dynex DSX and DS2 instruments. Deep well reservoir with 8 wells.
- Strip of eight wells
- Made of PP
Customers Who Bought This Also Bought