Plate, Round Well, J.T.Baker®
Supplier: Ritter, part of Avantor
Certificates
About this item
Made of virgin PP to minimise extractables.
- Raised rim to prevent cross contamination and enable a better heat-welded seal
- Also available rimless to allow insertion of vials, or when adhesive seals are used
- Round bottom for optimal mixing and recovery
- Suitable for use with automated sample handling systems
- Can be stored at –80 °C
J.T.Baker Plate Round well plus with air escapes and raised rims for unfreezing samples.
Specifications
- Catalog No:
- 43001-0010
- 43001-0016
- 43001-0116
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)