4-Way Collector (The Werner), SCAT
# de catalogue 76503-004
Fournisseur: SCAT AMERICAS, INC.
Spécifications
- Description:4-way collector (The Werner) for HPLC rinsing bottle GLS 80, PE-HD electrostatic conductive, 4× GL 45 (m)
- Cat. no.:76503-004
Spécifications
A propos de cet article
This purging manifold allows you to install up to four SafetyCaps on your purging bottle.
- Safe and easy handling - capillaries are reliably fixed
- Solvent filters have enough space due to the wide GLS 80 opening
- Simply close unused connections with standard bottle closures
The System for Solvent Safety stays permanently closed, the 'safety chain' remains intact during the purging process.
Customers Who Bought This Also Bought