SymLine® Tubes and Connectors, SCAT
Fournisseur: SCAT AMERICAS, INC.
Certificats
A propos de cet article
Place the collecting container/canister where needed; the pipe system can be exactly configured to suit specific needs.
- Straight pipes are available in standard lengths of 300, 900 and 1200 mm
- Angled pipes are available in the further lengths of 150 and 500 mm
- A wide range of connectors, branches, and manifolds allow the pipe system to be connected precisely to the conditions in the laboratory
The SymLine® pipe system provides the connection to the central collection point.
Customers Who Bought This Also Bought