SymLine® Grounding and Adapters
Fournisseur: SCAT AMERICAS, INC.
Certificats
A propos de cet article
Dovetail the SymLine® system to suit specific requirements. This product selection includes several useful ancillaries.
- SymLine® offers a suitable adapter for every common thread, to eliminate problems connecting different thread types
- Many useful accessories allow users to configure the SymLine® system according to specific needs
- All accessories are eclectically dissipative and approved for use in laboratories with explosion hazards
Dispose of liquid HPLC waste safely. SymLine® is quickly installed without drilling holes or making technical changes to existing laboratory equipment.
Customers Who Bought This Also Bought