SymLine® Safety Waste Caps and Ventilation Kit, SCAT
Fournisseur: SCAT AMERICAS, INC.
Certificats
A propos de cet article
Safety waste caps safely connect pipework to the waste container. The integrated exhaust filter blocks solvent vapors, for the protection of health and the environment.
- Caps are available with different thread sizes for attachment to the pipe system
- Caps are available with different numbers of hose connections for disposal of liquid waste
- By using a 4-in-1 collector, up to 4 safety waste caps can be connected simultaneously to only one connection point on the pipe system
- Ventilation tube, length 1500 mm, allows connection to lab exhaust system
- Adapter (Y-connector) for connecting two ventilation tubes to one common exhaust tube
Optionally, safety waste caps are available with electronic level sensors that indicate the degree of filling of the disposal container.
Customers Who Bought This Also Bought