Safety Funnels with Cap, SCAT
Fournisseur: SCAT AMERICAS, INC.
Certificats
A propos de cet article
Safety funnels with hinged lid cap, sieve and safety lance.
- Lid funnel with double thread R2" BSP/G2" (m) + 2" Tri-Sure (m) or S60/61
- Made of PE-HD material
- Stable stainless steel strainer
- Large funnel diameter: 250 mm
The funnels with hinged lid are suitable for universal use in the laboratory and are highly resistant to conventional chemicals due to the high-quality PE-Hd material.
Customers Who Bought This Also Bought