Multi-position Submersible Magnetic Stirrers, Crystal Industries
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
Certificates
About this item
These multi-position submersible magnetic stirrers feature a space saving design, ideal for stirring multiple size beakers or flasks simultaneously and have been constructed to operate safely in a water bath or CO₂ incubator.
- Stir a variety of beaker or flask sizes simultaneously in a compact space
- Waterproof body for submersible use in water baths
- Five stirring levels provide excellent mixing capability
- Sealed stainless steel body construction protects against intrusion when used in a CO₂ incubator
- 2-years replacement warranty
Multi-position magnetic stirrers are simple to use and save valuable bench space. Ideal for any laboratory that needs to stir a variety of beakers or flasks simultaneously. Constructed from high quality materials for safe, reliable operation in a water bath or CO₂ incubator.
Delivery: Stir bars included.
Customers Who Bought This Also Bought