Universal Waste Hubs, 'Jan', S.C.A.T.
Fournisseur: SCAT AMERICAS, INC.
Certificats
A propos de cet article
Collect your laboratory waste with our 'Jan' Universal Waste Hub, the smart all-in-one solution for liquid waste. It is a clever combination of lid funnel with tubing and HPLC connections plus an integrated exhaust air filter. Many other innovative details guarantee maximum safety compared to conventional solutions.
- High quality PE-HD and PTFE materials for maximum durability
- Stable funnel lid with integrated sealing lip against evaporation
- Removable dirt sieve for easy cleaning
- Funnel basin (140 mm) with high flow rate for larger disposal volumes
- Includes exhaust air filter size M with 6 months service life
- Accessories (fittings, connectors and blind plugs) already included
- Available as an electrically conductive version for flammable or explosive waste
The Universal Waste Hub 'Jan' is available in various thread sizes and can be expanded as required by means of adapters. It is also compatible with all SCAT exhaust air filter sizes (S/M/L).
For further details, please refer to the Related Information section below.
Customers Who Bought This Also Bought