VWR® M4 UV/Vis Spectrophotometer
Catalog # 76520-716
Supplier: VWR International
Specifications
- Model:M4
- Light source:Flashing xenon lamp
- Description:UV/Visible spectrophotometer
- Spectral bandwidth:2 nm
- Weight:10.5 kg
- Display:5" colour touch screen
- W×D×H:456×360×185 mm
- Wavelength range:190 - 1100 nm
- Output:USB; serial port; Bluetooth® (optional)
- Languages:6 can be switched: English, German, French, Spanish, Portuguese, simplified Chinese, Russian
- Cat. no.:76520-716
Specifications
About this item
The M4 single split-beam spectrophotometer, with flashing xenon lamp, is an easy to use, highly reliable and accurate product.
- Single split-beam design for sample and blank variability that can be monitored at the same time
- Xenon flash lamp with a very strong energy in the UV and visible range
- Connects to a computer based on Windows® through various interfaces
Measure absorbance and transmission at any defined wavelength, generates a graph and searches for peaks and valleys, also analyses the graph via built-in methods.
The application can be extended to luminosity determination, multiple wavelength measurement, quantitative analysis, dynamic determination, DNA/protein determination and spectral measurements.
Delivery: Supplied with a user manual, 10 mm quartz cuvette, 10 mm glass cuvette, fuse, power cable and dust cover.
Customers Who Bought This Also Bought