Accessories for Safety Funnels with Lid 'MARCO' V2.0
# de catalogue 76503-936
Fournisseur: SCAT AMERICAS, INC.
Spécifications
- Description:Replacement lid for funnel 'MARCO' V2.0
Spécifications
A propos de cet article
Replacement parts for 'MARCO' V2.0 safety funnels, which can be closed safely to prevent the danger of ignition.
- Suitable for all models of the MARCO series
- Available in conductive and non-conductive versions
- Stainless steel sieve to catch small objects such as stirrers from HPLC equipment
The 'MARCO' V2.0 safety funnel (sold separately) is a lid funnel for the disposal of liquid waste. The lid seals the funnel and prevents the escape of toxic or explosive vapours. Numerous features guarantee safe handling. Available in different thread sizes for safe attachment to laboratory bottles, canisters, or other containers.
Customers Who Bought This Also Bought