AmpliPur™ Expert Tips, Gilson
Fournisseur: GILSON, INC.
Certificats
A propos de cet article
AmpliPur™ Expert Tips are ISO 8655 compliant and provide a significant value for the money spent. They are equipped with a filter to help protect precious samples and your pipettes from cross-contamination.
- Designed to fit various pipette brands for maximum convenience on the bench
- Ideal for molecular biology
AmpliPur™ Expert Tips are certified DNase- and RNase-free. It makes them the preferred tool for molecular biology applications, supporting you in ensuring the limit of detection of your experiment, resulting in more sensitive nucleic acid quantitation.
These tips significantly contribute to molecular testing performance by ensuring sample integrity during the pipetting process.
Certifications: Manufactured according to the certified Gilson quality standard, AmpliPur Expert Tips comply with ISO 8655 requirements and are certified free of RNase and DNase, preserving your sample integrity.
Customers Who Bought This Also Bought