VWR® Metal Dissecting Scalpel
Fournisseur: VWR International
Certificats
A propos de cet article
VWR® Standard Metal Scalpel is perfect for dissecting applications.
- Easy to clean and sterilize
- Reusable
- Smooth, antiglare finish handle
- Useful in educational, research or healthcare setting
- Fully autoclavable
Nickel-plated Scalpels.
Certifications: ISO, GMP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.