VWR® Scalpel Knife Handle with Graduated Measurements
Supplier: VWR International
Certificates
About this item
These high-quality scalpel knife handle fits the VWR® 10 and 20 series scalpel blades.
- Scale for measurements
- Secure blade fitment
- Autoclavable/sterilizable
- Ribbed/grooved
For classroom dissection, field-study, removal of small embedded objects and crafts.
Certifications: ISO, GMP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.