VWR® Jar Forceps with Serrated Tips
Fournisseur: VWR International
Certificats
A propos de cet article
These high-quality thumb forceps remove specimens from deep bottles or jars.
- Manufactured from AISI 400 series stainless steel
- High polish, mirror finish
- Autoclavable and corrosion-resistant
For classroom dissecting, field-study, removal of small embedded objects and crafts.
Certifications: ISO, GMP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.