Safety Caps and Safety Waste Cap Accessories and Replacement Parts, S.C.A.T.
Fournisseur: SCAT AMERICAS, INC.
Certificats
A propos de cet article
Helpful supplies for the safe use of SCAT and SymLine products safety caps and safety waste caps.
- SCAT connectors are available in the following versions:
- Conical connectors
- Y-connectors
- Tube connectors
- Blind plugs
- Capillary connector
SCAT thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters, or other disposal and supply units.
Customers Who Bought This Also Bought